Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hydroxymethyl)benzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39434-0010 39434-0100
TS39434-0010EA 63 USD InStock
TS39434-0010 TS39434-0100
4-(Hydroxymethyl)benzoic acid 98%
4-(Hydroxymethyl)benzoic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00017598
CAS Number: 3006-96-0
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 181 to 186 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty