Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)benzenesulphonyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39776-0010 39776-0050
TS39776-0010EA 28 USD InStock
TS39776-0010 TS39776-0050
4-(Trifluoromethyl)benzenesulphonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride


Formula: C₇H₄ClF₃O₂S
MW: 244.62
Boiling Pt: 118…120 °C (18 mmHg)
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty