Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl hydrogen isophthalate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

39823-0010 39823-0050
TS39823-0010EA 45 USD InStock
TS39823-0010 TS39823-0050
Methyl hydrogen isophthalate 97%
Methyl hydrogen isophthalate


Formula: C₉H₈O₄
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00029972
CAS Number: 1877-71-0
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 191 to 197 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty