Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-difluorobenzoic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39904-0050 39904-0250
TS39904-0050EA 85 USD InStock
TS39904-0050 TS39904-0250
2-Bromo-4,5-difluorobenzoic acid 97%
2-Bromo-4,5-difluorobenzoic acid


Formula: C₇H₃BrF₂O₂
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD04973422
CAS Number: 64695-84-7
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 122 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty