Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-hydroxy-4-methoxybenzoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39926-0010 39926-0050
TS39926-0010EA 45 USD InStock
TS39926-0010 TS39926-0050
Methyl-3-hydroxy-4-methoxybenzoate 98%
Methyl-3-hydroxy-4-methoxybenzoate


Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01321262
CAS Number: 6702-50-7
Order Now
Specification Test Results
Appearance Off-white to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 62 to 67 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty