Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dimethylbenzophenone 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

40822-0010 40822-0100
TS40822-0010EA 15 USD InStock
TS40822-0010 TS40822-0100
4,4'-Dimethylbenzophenone 98+%
4,4'-Dimethylbenzophenone


Formula: C₁₅H₁₄O
Boiling Pt: 200 °C (17 mmHg)
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00017214
CAS Number: 611-97-2
EINECS: 210-287-6
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 93 to 97 °C
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty