Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-p-cresol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41385-0250 41385-1000
TS41385-0250EA 55 USD InStock
TS41385-0250 TS41385-1000
2-Methoxy-p-cresol 99%
2-Methoxy-p-cresol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 219…222 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.09 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 13,02599
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid (May turn yellow on storage)
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5400 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty