Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TCSA 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42023-0050 42023-0250
TS42023-0050EA 129 USD InStock
TS42023-0050 TS42023-0250
TCSA 99%
TCSA


Formula: C₁₃H₇Cl₄NO₂
Melting Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00041745
CAS Number: 1154-59-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥98.5 %
HPLC ≥94 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty