Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-methylbenzaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42702-0010 42702-0050
TS42702-0010EA 50 USD InStock
TS42702-0010 TS42702-0050
4-Methoxy-2-methylbenzaldehyde 97%
4-Methoxy-2-methylbenzaldehyde


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02261771
CAS Number: 52289-54-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty