Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43032-0010 43032-0050
TS43032-0010EA 173 USD InStock
TS43032-0010 TS43032-0050
3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%
3-Fluoro-5-(trifluoromethyl)phenylboronic acid


Formula: C₇H₅BF₄O₂
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD06657735
CAS Number: 159020-59-4
Order Now
Specification Test Results
Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 175 to 180 °C
HPLC ≥96.0 %
Substance darkened by H2SO4 Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty