Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(1,1-difluoroethyl)benzene 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43253-0050 43253-0100
TS43253-0050EA 304 USD InStock
TS43253-0050 TS43253-0100
1-Bromo-4-(1,1-difluoroethyl)benzene 95%
1-Bromo-4-(1,1-difluoroethyl)benzene


Formula: C₈H₇BrF₂
MW: 221.05 g/mol
Boiling Pt: 80…82 °C (10Torr)
Storage Temperature: Ambient
MDL Number: MFCD11110333
CAS Number: 1000994-95-5
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Density 1.76 to 1.80 g/ml (25°C)
Additional info dielectric strength: ≥35 kV
Viscosity 0.7 to 0.9 cSt (at 25°C)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty