Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxybenzonitrile 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43316-0050 43316-0250
TS43316-0050EA 45 USD InStock
TS43316-0050 TS43316-0250
2-Methoxybenzonitrile 99%
2-Methoxybenzonitrile


Formula: C₈H₇NO
Boiling Pt: 256 °C (760 mmHg)
Melting Pt: 21…22 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001783
CAS Number: 6609-56-9
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.0 %
Residue after evaporation ≤0.05 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty