Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chlorophenyl)acetylene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43475-0010 43475-0050
TS43475-0010EA 85 USD InStock
TS43475-0010 TS43475-0050
(2-Chlorophenyl)acetylene 98%
(2-Chlorophenyl)acetylene


Formula: C₈H₅Cl
Boiling Pt: 82 °C (16hPa)
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269951
CAS Number: 873-31-4
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC 97.5 %
Titration with HClO4 97.5 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty