Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-3-pyrroline 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43677-0010 43677-0050
TS43677-0010EA 38 USD InStock
TS43677-0010 TS43677-0050
1-Benzyl-3-pyrroline 97%
1-Benzyl-3-pyrroline


Formula: C₁₁H₁₃N
Boiling Pt: 92 °C (1hPa)
Density: 0.837 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00012324
CAS Number: 6913-92-4
UN: 3267
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty