Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2-phenylpropanenitrile 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43825-0010 43825-0050
TS43825-0010EA 91 USD InStock
TS43825-0010 TS43825-0050
2-Methyl-2-phenylpropanenitrile 97%
2-Methyl-2-phenylpropanenitrile


Formula: C₁₀H₁₁N
Boiling Pt: 54…56 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00099614
CAS Number: 1195-98-8
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty