Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorophenylboronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44469-0050 44469-0250
TS44469-0050EA 77 USD InStock
TS44469-0050 TS44469-0250
4-Chlorophenylboronic acid pinacol ester 97%
4-Chlorophenylboronic acid pinacol ester


Formula: C₁₂H₁₆BClO₂
MW: 238.52
Melting Pt: 55…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD05663875
CAS Number: 195062-61-4
Order Now
Specification Test Results
Appearance White to off-white Solid or crystalline solid
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty