Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45094-0010 45094-0050
TS45094-0010EA 150 USD InStock
TS45094-0010 TS45094-0050
6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
6-Hydroxybenzo[d]thiazole-2-carbonitrile


Formula: C₈H₄N₂OS
Melting Pt: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00296905
CAS Number: 939-69-5
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty