Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chloroethyl)-2-pyrrolidone 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45376-0010 45376-0050
TS45376-0010EA 87 USD InStock
TS45376-0010 TS45376-0050
1-(2-Chloroethyl)-2-pyrrolidone 95%
1-(2-Chloroethyl)-2-pyrrolidone


Formula: C₆H₁₀ClNO
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00169064
CAS Number: 51333-90-5
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty