Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Bis(trifluoromethyl)benzidine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45757-0050 45757-0250
TS45757-0050EA 163 USD InStock
TS45757-0050 TS45757-0250
2,2'-Bis(trifluoromethyl)benzidine 97%
2,2'-Bis(trifluoromethyl)benzidine


Formula: C₁₄H₁₀F₆N₂
MW: 320.24 g/mol
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00190155
CAS Number: 341-58-2
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Melting point 180 to 186 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty