Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Ethyl-10-hydroxycamptothecin 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45909-0010 45909-1000
TS45909-0010EA 366 USD InStock
TS45909-0010 TS45909-1000
7-Ethyl-10-hydroxycamptothecin 98%
7-Ethyl-10-hydroxycamptothecin


Formula: C₂₂H₂₀N₂O₅
MW: 392.40 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00871873
CAS Number: 86639-52-3
UN: 1544
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty