Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-fluorobenzoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45924-0250 45924-1000
TS45924-0250EA 70 USD InStock
TS45924-0250 TS45924-1000
Methyl-4-fluorobenzoate 98%
Methyl-4-fluorobenzoate


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 90…92 °C (27hPa)
Density: 1.192 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00017959
CAS Number: 403-33-8
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty