Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-isothiazolin-3-one 50% in aqueous solution

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45997-0050 45997-0250
TS45997-0050EA 65 USD InStock
TS45997-0050 TS45997-0250
2-Methyl-4-isothiazolin-3-one 50% in aqueous solution
2-Methyl-4-isothiazolin-3-one


Formula: C₄H₅NOS
Density: 1.16 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD01742315
CAS Number: 2682-20-4
UN: 2927
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
pH 5.0 to 7.0 (1 % in water)
Concentration ≥50 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty