Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromoaniline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46056-0050 46056-0250
TS46056-0050EA 96 USD InStock
TS46056-0050 TS46056-0250
3,5-Dibromoaniline 98%
3,5-Dibromoaniline


Formula: C₆H₅Br₂N
Melting Pt: 53…54.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00047841
CAS Number: 626-40-4
Order Now
Specification Test Results
Appearance Brown to black Solid or crystalline solid
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty