Evergreen Funnels, Caplugs Evergreen
Supplier: Caplugs
208-5122-010 208-5136-030
76521-498CT 45.76 USD InStock
76521-498 76521-500
Evergreen Funnels, Caplugs Evergreen
Funnels Filtration Funnels Filter Funnels
Caplugs Evergreen funnels feature internal ribs to promote faster flow and external ribs to help prevent airlock.
  • Polystyrene funnels are appropriate for use with mild acidic or alkali solutions
  • Polypropylene funnels are suitable for use with organic solvents and stronger acids and alkalis
Funnels measure approximately 60 mm (2½") wide at the top with a 60° taper to the funnel stem.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty