Eisco Narrow Mouth Polypropylene Reagent Bottles
Supplier: Wards
CH0172CN CH0170CN CH0170BN CH0170ANPK10 CH0172EN CH0172BN CH0170AN CH0170DN CH0172AN CH0172DN
470343-564EA 3.09 USD InStock
470343-564 470343-940 470343-996 470344-044 470344-190 470344-316 470344-348 470344-444 470345-204 470345-540
Eisco Narrow Mouth Polypropylene Reagent Bottles
Bottles Plastic Bottles
Ideal for holding small liquid or powder reagents.
  • Industrial quality reagent bottle
  • Excellent for storing chemicals, liquids or powders
  • Rigid polypropylene construction provides excellent chemical resistance to acids, alcohols and alkalis, and very good resistance to aldehydes and hydrocarbons
  • Both bottle and cap are translucent with excellent optical clarity
Narrow neck reagent bottle with leak-proof screw cap. Stable at temperatures between −20 to +135 °C. Bottle and cap are both autoclavable and top rack dishwasher safe.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty