Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Azidopropionic acid 95%

Supplier: AFG BIOSCIENCE LLC

C56674-1G C56674-5G
76620-288EA 98.58 USD InStock
76620-288 76620-290
3-Azidopropionic acid 95%
3-Azidopropionic acid


Formula: C₃H₅N₃O₂
MW: 115.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01695706
CAS Number: 18523-47-2
Order Now
Specification Test Results
Purity Limit 95%
Appearance Yellow Oil

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty