Anti-Cytokeratin AE Acidic Mouse Monoclonal Antibody [clone: AE1]
Supplier: Diagnostic Biosystems
MOB092-01 MOB092-05 MOB092
76633-274EA 115.9 USD InStock
76633-274 76633-276 76633-278
Anti-Cytokeratin AE Acidic Mouse Monoclonal Antibody [clone: AE1]
Antibodies
This antibody recognizes most of the acidic (Type I) keratins. It recognizes keratin polypeptides of 50 and 56.5 kDa. In keratinized epidermis, 50 kDa keratin is present in the basal layer, while 56.5 kDa keratin is present in suprabasal layers.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty