Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Supplier: Diagnostic Biosystems
MOB098-01 MOB098-05 MOB098
76633-292EA 119.72 USD InStock
76633-292 76633-294 76633-296
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Antibodies
This antibody reacts with a glycoprotein of MW 110 kDa, present on the surface of hematopoietic cells i.e. myeloid and lymphoid progenitors. It stains the vascular endothelial cells.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty