Anti-Chromogranin A Mouse Monoclonal Antibody [clone: LK2H10]
Supplier: Diagnostic Biosystems
MOB048-01 MOB048-05 MOB048
76633-146EA 154.11 USD InStock
76633-146 76633-148 76633-150
Anti-Chromogranin A Mouse Monoclonal Antibody [clone: LK2H10]
Antibodies
Chromogranin A is a 439 amino acid protein present in secretory granules of a wide variety of endocrine cells and neurons. A positive staining is seen in the secretory granules of parathyroid, adrenal medulla, anterior pituitary gland and Langerhans islets of pancreas. This antibody stains a variety of neuroendocrine tumors.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty