Anti-Timp-2 Mouse Monoclonal Antibody [clone: 3A4]
Supplier: Diagnostic Biosystems
MOB318-01 MOB318-05 MOB318
76633-656EA 138.83 USD InStock
76633-656 76633-658 76633-660
Anti-Timp-2 Mouse Monoclonal Antibody [clone: 3A4]
Antibodies
This antibody is specific to 21 kDa protein known as TIMP-2. TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family. TIMP are of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (MMP-2) and active form of 92 kDa type IV collagenase (MMP-9). TIMP inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty