Anti-Synaptophysin (SYP02) Mouse Monoclonal Antibody [clone: SYP02]
Supplier: Diagnostic Biosystems
MOB399-01 MOB399-05 MOB399
76633-808EA 108.26 USD InStock
76633-808 76633-810 76633-812
Anti-Synaptophysin (SYP02) Mouse Monoclonal Antibody [clone: SYP02]
Antibodies
This antibody is specific to a 38 kDa protein. Synaptophysin is a glycoprotein occurring in presynaptic vesicles of neurons in the brain, spinal cord, retina, vesicles of adrenal medulla and in neuromuscular junctions. This antibody reacts with neuroendocrine neoplasms of neural as well as epithelial types.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty