Anti-CD3 Mouse Monoclonal Antibody [clone: LN10]
Supplier: Diagnostic Biosystems
MOB474-01 MOB474-05 MOB474
76634-000EA 142.64 USD InStock
76634-000 76634-002 76634-004
Anti-CD3 Mouse Monoclonal Antibody [clone: LN10]
Antibodies
The clone F7.2.38 is specific for the non-glycosylated epsilon chain of the human CD3 molecule. Clone F7.2.38 recognizes T cells in the thymus, bone marrow, peripheral lymphoid tissue and blood and is a pan T cell marker. The CD3 molecule consists of five different polypeptide chains with molecular weights ranging from 16 to 28 kDa. The CD3 antigen is first detected in early thymocytes, and its appearance probably represents one of the earliest signs of commitment to the T cell lineage.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty