Anti-INSM1 (A-8) Mouse Monoclonal Antibody [clone: A-8]
Supplier: Diagnostic Biosystems
MOB586-01 MOB586-05 MOB586
76634-198EA 96.8 USD InStock
76634-198 76634-200 76634-202
Anti-INSM1 (A-8) Mouse Monoclonal Antibody [clone: A-8]
Antibodies
INSM1 (insulinoma-associated protein 1), also known as zinc-finger protein IA-1, is a developmentally regulated zinc-finger transcription factor. It localizes to the nucleus and is expressed in embryonic tissues undergoing neuroendocrine differentiation. INSM1 is not expressed in normal adult tissues but it can be found highly expressed in neuroendocrine tumors. INSM1 contains five Cys2-His2-type zinc-finger DNA binding domains and a prohormone domain. INSM1 acts as a transcriptional repressor of the Neuro D promoter and recruits cyclin D1 as a corepressor. It plays an important role in neuroendocrine development and is required for normal differentiation of pancreatic endocrine cells. Inhibition of INSM1 results in decreased formation of glucagon and Insulin positive cells. The gene encoding INSM1 is directly regulated by Neurogenin 3 which binds chromatin in the INSM1 promoter region and induces transcription.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty