Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin, granular 0.1 --> 0.8 mm

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22257-1000 222575000
TS22257-1000EA 52 USD InStock
TS22257-1000 TS222575000
Tin, granular 0.1 --> 0.8 mm
Tin


Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587
Order Now
Specification Test Results
Appearance Grey Granular powder
Particle size 0.1 to 0.8 mm
Antimony (Sb) ≤200 ppm
Bismuth (Bi) ≤200 ppm
Copper (Cu) ≤100 ppm
Iron (Fe) ≤500 ppm
Lead (Pb) ≤700 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty