Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraphenylphosphonium chloride hydrate 99%

Supplier: AMBEED, INC
Warning

A1002319-1000G A1002319-100G A1002319-25G A1002319-500G
76645-932EA 57 USD InStock
76645-932 76645-934 76645-936 76645-938
Tetraphenylphosphonium chloride hydrate 99%
Tetraphenylphosphonium chloride hydrate
Precaución: for Research Use Only


Formula: C₂₄H₂₂ClOP
CAS Number: 123557-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty