Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-chloro-5-methoxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A1002571-1G A1002571-250MG A1002571-5G
76645-948EA 179 USD InStock
76645-948 76645-950 76645-952
2-Amino-4-chloro-5-methoxybenzonitrile 97%
2-Amino-4-chloro-5-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₇ClN₂O
MDL Number: MFCD28131187
CAS Number: 1824059-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty