Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-4-oxo-1,4-dihydropyridine-2-carboxylic acid 96% (dried basis) may cont. up to 8% water

Supplier: AMBEED, INC
Warning

A1002949-1G A1002949-250MG
76645-986EA 82 USD InStock
76645-986 76645-988
5-(Benzyloxy)-4-oxo-1,4-dihydropyridine-2-carboxylic acid 96% (dried basis) may cont. up to 8% water
5-(Benzyloxy)-4-oxo-1,4-dihydropyridine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₁NO₄
MDL Number: MFCD18446980
CAS Number: 107550-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty