Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carbamoyl-1-methylpyridin-1-ium iodide 97%

Supplier: AMBEED, INC
Warning

A1003172-1G A1003172-5G
76646-018EA 62 USD InStock
76646-018 77619-496
3-Carbamoyl-1-methylpyridin-1-ium iodide 97%
3-Carbamoyl-1-methylpyridin-1-ium iodide
Precaución: for Research Use Only


Formula: C₇H₉IN₂O
MW: 264.0636 g/mol
MDL Number: MFCD00031898
CAS Number: 6456-44-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty