Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(3-bromophenyl)phosphine Oxide 98%

Supplier: AMBEED, INC
Warning

A1003229-100MG A1003229-1G A1003229-250MG
76646-050EA 16 USD InStock
76646-050 76646-052 76646-054
Tris(3-bromophenyl)phosphine Oxide 98%
Tris(3-bromophenyl)phosphine Oxide
Precaución: for Research Use Only


Formula: C₁₈H₁₂Br₃OP
MDL Number: MFCD32062937
CAS Number: 38019-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty