Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromo-4-isopropylphenoxy)-N-(2,2,6,6-tetramethylpiperidin-4-yl)acetamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1324873-10MG A1324873-1MG A1324873-50MG A1324873-5MG
76657-866EA 52 USD InStock
76657-866 76657-868 76657-870 76657-872
2-(2-Bromo-4-isopropylphenoxy)-N-(2,2,6,6-tetramethylpiperidin-4-yl)acetamide hydrochloride 97%
2-(2-Bromo-4-isopropylphenoxy)-N-(2,2,6,6-tetramethylpiperidin-4-yl)acetamide hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₃₂BrClN₂O₂
MDL Number: MFCD05985784
CAS Number: 1052515-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty