Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Dodecyl-3-methylimidazolium bromide 98%

Supplier: AMBEED, INC
Warning

A1003360-25G A1003360-5G
76646-128EA 30 USD InStock
76646-128 76646-130
1-Dodecyl-3-methylimidazolium bromide 98%
1-Dodecyl-3-methylimidazolium bromide
Precaución: for Research Use Only


Formula: C₁₆H₃₁BrN₂
MDL Number: MFCD17014729
CAS Number: 61546-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty