Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nitazoxanide 97%

Supplier: AMBEED, INC
Warning

A1003469-1000G A1003469-100G A1003469-10G A1003469-25G A1003469-500G
76646-158EA 397 USD InStock
76646-158 76646-160 76646-162 76646-164 76646-166
Nitazoxanide 97%
Nitazoxanide
Precaución: for Research Use Only


Formula: C₁₂H₉N₃O₅S
MDL Number: MFCD00416599
CAS Number: 55981-09-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty