Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%

Supplier: AMBEED, INC
Warning

A1003828-100MG A1003828-1G A1003828-250MG
76646-214EA 96 USD InStock
76646-214 76646-216 76646-218
(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%
(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₃₀Cl₄N₄
MDL Number: MFCD16621417
CAS Number: 1285550-13-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty