Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1-Trifluoro-N-[(11bR)-4-oxido-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl]methanesulfonamide 97%

Supplier: AMBEED, INC
Warning

A1005266-100MG A1005266-1G A1005266-250MG A1005266-50MG
76646-368EA 184 USD InStock
76646-368 76646-370 76646-372 76646-374
1,1,1-Trifluoro-N-[(11bR)-4-oxido-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl]methanesulfonamide 97%
1,1,1-Trifluoro-N-[(11bR)-4-oxido-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl]methanesulfonamide
Precaución: for Research Use Only


Formula: C₃₃H₂₁F₃NO₅PS
CAS Number: 1010799-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty