Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-2-methylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A1005886-1G A1005886-250MG A1005886-5G
76646-418EA 12 USD InStock
76646-418 76646-420 76646-422
4-Acetyl-2-methylbenzoic acid 97%
4-Acetyl-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
MDL Number: MFCD20639525
CAS Number: 55860-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty