Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R,R)-iPr-BI-DIME 97%

Supplier: AMBEED, INC
Warning

A1005934-100MG A1005934-250MG A1005934-50MG
76646-440EA 105 USD InStock
76646-440 76646-442 77619-582
(R,R)-iPr-BI-DIME 97%
(R,R)-iPr-BI-DIME
Precaución: for Research Use Only


Formula: C₂₂H₂₉O₃P
MDL Number: MFCD31630762
CAS Number: 1477517-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty