Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11bS)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%

Supplier: AMBEED, INC
Warning

A1006121-100MG A1006121-1G A1006121-250MG
76646-580EA 50 USD InStock
76646-580 76646-582 76646-584
(11bS)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bS)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only


Formula: C₃₂H₁₉Cl₂O₄P
MW: 569.3706 g/mol
CAS Number: 1945966-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty