Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[(3-Bromophenyl)methyl]hydrazine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A1012220-100MG A1012220-250MG A1012220-1G
76646-726EA 29 USD InStock
76646-726 76646-728 76671-046
[(3-Bromophenyl)methyl]hydrazine hydrochloride 95%
[(3-Bromophenyl)methyl]hydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₀BrClN₂
MDL Number: MFCD16660424
CAS Number: 1384427-52-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty