Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5H,6H,7H-Pyrazolo[3,2-b][1,3]oxazin-6-ol 95%

Supplier: AMBEED, INC
Warning

A1014753-100MG A1014753-50MG
76646-736EA 253 USD InStock
76646-736 76646-738
5H,6H,7H-Pyrazolo[3,2-b][1,3]oxazin-6-ol 95%
5H,6H,7H-Pyrazolo[3,2-b][1,3]oxazin-6-ol
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
MDL Number: MFCD28639709
CAS Number: 1558277-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty