Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1H-benzotriazole (mixture) 98%

Supplier: AMBEED, INC
Warning

A123632-1000G A123632-100G A123632-25G A123632-500G
76646-900EA 43 USD InStock
76646-900 76646-902 76646-904 76646-906
Methyl-1H-benzotriazole (mixture) 98%
Methyl-1H-benzotriazole (mixture)
Precaución: for Research Use Only


Formula: C₇H₇N₃
MDL Number: MFCD00167158
CAS Number: 29385-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty